missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EML4, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB31053.100uL
This item is not returnable.
View return policy
Description
Sequence: MSIIQWKLVEKLSLPQNETVADTTLTKAPVSSTESVIQSNTPTPPPSQPLNETAEEESRISSSPTLLENSLEQTVEPSEDHSEEESSpecifications
| EML4 | |
| Polyclonal | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000-2500) Western Blot (1:100-250) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| echinoderm microtubule associated protein like 4 | |
| EML4 | |
| Recombinant protein corresponding to human EML4. | |
| RUO | |
| 27436 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
| C2orf2/DKFZp686P18118/ELP120/FLJ10942/FLJ32318/ROPP120 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction