missing translation for 'onlineSavingsMsg'
Learn More

EML2 Antibody (2H6), Novus Biologicals™

Product Code. 18398829 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18398829 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18398829 Supplier Novus Biologicals Supplier No. H00024139M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

EML2 Monoclonal antibody specifically detects EML2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen EML2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA
Classification Monoclonal
Clone 2H6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH32630
Gene Alias echinoderm microtubule associated protein like 2, echinoderm microtubule-associated protein-like 2, echinoderm MT-associaated protein (EMAP)-like protein 70, echinoderm MT-associated protein (EMAP)-like protein 70, ELP70, EMAP-2huEMAP-2, EMAP2microtubule-associated protein like echinoderm EMAP, EMAPL2, HuEMAP-2, TRTLT2
Host Species Mouse
Immunogen EML2 (AAH32630, 1 a.a. ~ 427 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSSFGAGKTKEVIFSVEDGSVKMFLRGRPVPMMIPDELAPTYSLDTRSELPSCRLKLEWVYGYRGRDCRANLYLLPTGEIVYFVASVAVLYSVEEQRQRHYLGHNDDIKCLAIHPDMVTIATGQVAGTTKEGKPLPPHVRIWDSVSLSTLHVLGLGVFDRAVCCVGFSKSNGGNLLCAVDESNDHMLSVWDWAKETKVVDVKCSNEAVLVATFHPTDPTVLITCGKSHIYFWTLEGGSLSKRQGLFEKHEKPKYVLCVTFLEGGDVVTGDSGGNLYVWGKGGNRITQAVLGAHDGGVFGLCALRDGTLVSGGGRDRRVVLWGSDYSKLQEVEVPEDFGPVRTVAEGHGDTLYVGTTRNSILQGSVHTGFSLLVQGHVEELWGLATHPSRAQFVTCGQDKLVHLWSSDSHQPLWSRIIEMAAAGHGDP
Purification Method IgG purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 24139
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.