missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EML1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111287
This item is not returnable.
View return policy
Description
Immunogen sequence: SGVRKETAVP ATKSNIKRTS SSERVSPGGR RESNGDSRGN RNRTGSTSSS Highest antigen sequence indentity to the following orthologs: Rat - 82%, Mouse - 82%.
Human echinoderm microtubule-associated protein-like is a strong candidate for the Usher syndrome type 1A gene. Usher syndromes (USHs) are a group of genetic disorders consisting of congenital deafness, retinitis pigmentosa, and vestibular dysfunction of variable onset and severity depending on the genetic type. The disease process in USHs involves the entire brain and is not limited to the posterior fossa or auditory and visual systems. The USHs are categorized as type I (USH1A, USH1B, USH1C, USH1D, USH1E and USH1F), type II (USH2A and USH2B) and type III (USH3). The type I is the most severe form. Gene loci responsible for these three types are all mapped. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| EML1 | |
| Polyclonal | |
| Unconjugated | |
| EML1 | |
| 1110008N23Rik; A930030P13Rik; AA171013; AI847476; AI853955; echinoderm microtubule associated protein like 1; echinoderm microtubule associated protein-like protein 1; echinoderm microtubule-associated protein-like 1; ELP79; EMAP; EMAP1; EMAP-1; EMAPL; EMAPL1; Eml1; HuEMAP; huEMAP-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2009 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00423 | |
| EML1 | |
| Recombinant Protein corresonding to Human EML1. Recombinant protein control fragment (Product #RP-103045). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction