missing translation for 'onlineSavingsMsg'
Learn More

EML 425, MedChemExpress

Product Code. 16464368
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16464368

Brand: Medchem Express HY110263

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress EML425 is a potent and selective CREB binding protein (CBP)/p300 inhibitor with IC50s of 2.9 and 1.1 μM, respectively.

EML425 is a potent and selective CREB binding protein (CBP)/p300 inhibitor with IC50s of 2.9 and 1.1 μM, respectively.

Specifications

Chemical Name or Material EML 425
CAS 1675821-32-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H24N2O4
Quantity 10 mg
Solubility Information DMSO : ≥ 250 mg/mL (567.55 mM)
SMILES O=C(N(C(N(CC1=CC=CC=C1)C/2=O)=O)CC3=CC=CC=C3)C2=C/C4=C(C=C(O)C=C4C)C
Molecular Weight (g/mol) 440.49
Formula Weight 440.49
Percent Purity 98.16%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.