missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ EML 425
Description
EML 425 is a reversible and non-competitive CBP/p300 inhibitor (IC50 values are 1.1 and 2.9 μM, respectively). Cell permeable.
Specifications
Specifications
| Chemical Name or Material | 5-[(4-Hydroxy-2,6-dimethylphenyl)methylene]-1,3-bis(phenylmethyl)-2,4,6(1H,3H,5H)-pyrimidinetrione |
| CAS | 1675821-32-5 |
| Quantity | 10 mg |
| Product Line | Tocris |
| Target | Histone Acetyltransferase Inhibitors |
| Molecular Formula | C27H24N2O4 |
| Formula Weight | 440.49 |
| Purity | >98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?