missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ EML 425
GREENER_CHOICE

Product Code. p-7110859
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18754448 10 mg 10mg
18721588 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18754448 Supplier Tocris Bioscience™ Supplier No. 5646/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reversible and non-competitive CBP/p300 inhibitor

EML 425 is a reversible and non-competitive CBP/p300 inhibitor (IC50 values are 1.1 and 2.9 μM, respectively). Cell permeable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-[(4-Hydroxy-2,6-dimethylphenyl)methylene]-1,3-bis(phenylmethyl)-2,4,6(1H,3H,5H)-pyrimidinetrione
CAS 1675821-32-5
Quantity 10 mg
Target Histone Acetyltransferase Inhibitors
Molecular Formula C27H24N2O4
Formula Weight 440.49
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.