missing translation for 'onlineSavingsMsg'
Learn More

Emixustat, MedChemExpress

Product Code. p-200101210
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493807 50 mg 50mg
16487676 100 mg 100mg
16450328 5 mg 5mg
16430326 2 mg 2mg
16417868 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493807 Supplier Medchem Express Supplier No. HY19720

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Emixustat, a novel visual cycle modulator, is an inhibitor of the visual cycle isomerase with an IC50 value of 4.4 nM in vitro.

Emixustat, a novel visual cycle modulator, is an inhibitor of the visual cycle isomerase with an IC50 value of 4.4 nM in vitro.

Specifications

Chemical Name or Material Emixustat
CAS 1141777-14-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H25NO2
Quantity 50 mg
Synonym ACU-4429
Solubility Information Ethanol : 100 mg/mL (379.68 mM; Need ultrasonic) ∣DMSO : ≥ 43 mg/mL (163.26 mM)
SMILES O[C@H](CCN)C1=CC(OCC2CCCCC2)=CC=C1
Molecular Weight (g/mol) 263.38
Formula Weight 263.38
Percent Purity 98.74%
Grade Research
Physical Form Oil
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.