missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emixustat hydrochloride, MedChemExpress
Description
Emixustat hydrochloride strongly inhibits 11-cis-retinol production with IC50 values of 232 (plusmn) 3 nM.
Specifications
Specifications
| Chemical Name or Material | Emixustat hydrochloride |
| CAS | 1141934-97-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C16H26ClNO2 |
| Quantity | 100 mg |
| Synonym | ACU-4429 hydrochloride |
| Solubility Information | DMSO : ≥ 44 mg/mL (146.74 mM) |
| SMILES | O[C@H](CCN)C1=CC(OCC2CCCCC2)=CC=C1.[H]Cl |
| Molecular Weight (g/mol) | 299.84 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?