Learn More
Invitrogen™ EMILIN3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140216
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LLEEYVDRRL HRLWGSLLDG FEQKLQGVQS ECDLRVQEVR RQCEEGQAAS RRLHQSLDGR ELALRQELSQ LGSQLQGLSV SGRGSCCGQL ALINARMDG Highest antigen sequence indentity to the following orthologs: Rat - 85%, Mouse - 85%.
Specifications
| EMILIN3 | |
| Polyclonal | |
| Unconjugated | |
| EMILIN3 | |
| C20orf130; dJ620E11.4; Elastin microfibril interface-located protein 3; elastin microfibril interface-located protein 5; Elastin microfibril interfacer 3; elastin microfibril interfacer 5; EMILIN3; EMILIN-3; EMILIN5; EMILIN-5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90187 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NT22 | |
| EMILIN3 | |
| Recombinant protein corresponding to Human EMILIN3. Recombinant protein control fragment (Product #RP-109679). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.