missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMILIN1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA551745
This item is not returnable.
View return policy
Description
Immunogen sequence: PQSIMYRRFL RPRYRVAYKT VTDMEWRCCQ GYGGDDCAES PAPALGPASS TPRPLARPAR PNLSGSSAGS PLSGLGGEGP GESEKVQQLE EQVQSLTKEL QGLRGVLQGL SGRLAEDVQR AVETAFNGRQ QPADAAARPG VHETLNE Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 74%.
This gene encodes a protein that is one of the two components of elastic fibers. The encoded protein is rich in hydrophobic amino acids such as glycine and proline, which form mobile hydrophobic regions bounded by crosslinks between lysine residues. Deletions and mutations in this gene are associated with supravalvular aortic stenosis (SVAS) and autosomal dominant cutis laxa. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
EMILIN1 | |
Polyclonal | |
Unconjugated | |
EMILIN1 | |
5830419M17Rik; AW229038; BB105748; elastin microfibril interface located protein 1; elastin microfibril interface-located protein 1; Elastin microfibril interfacer 1; EMI; EMILIN; EMILIN1; EMILIN-1; gp115; LOW QUALITY PROTEIN: EMILIN-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11117 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.11 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y6C2 | |
EMILIN1 | |
Recombinant protein corresponding to Human EMILIN1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only