missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMI1 Antibody (EMI1/1176) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-48221
This item is not returnable.
View return policy
Description
EMI1 Monoclonal specifically detects EMI1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| EMI1 | |
| Monoclonal | |
| Unconjugated | |
| 1.0mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| FBXO5 | |
| Recombinant fragment (203 amino acid residues between aa 1-250) of human EMI1 protein (Uniprot: Q9UKT4) | |
| Protein A or G purified | |
| RUO | |
| 26271 | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| EMI1/1176 | |
| Immunohistochemistry-Paraffin | |
| EMI1Early mitotic inhibitor 1, F-box only protein 5, F-box protein 5, FBX5F-box protein Fbx5, Fbxo31 | |
| Mouse | |
| 56 kDa | |
| 7 mL | |
| Primary | |
| It recognizes a 56kDa protein, which is identified as Early Mitotic Inhibitor-1 (EMI1). It regulates mitosis by inhibiting the anaphase promoting complex/cyclosome (APC). Emi1 is a conserved F box protein containing a zinc-binding region essential for APC inhibition. The Emi1 protein functions to promote cyclin A accumulation and S phase entry in somatic cells by inhibiting the APC complex. At the G1-S transition, Emi1 is transcriptionally induced by the E2F transcription factor. Emi1 overexpression accelerates S-phase entry and can override a G1 block caused by overexpression of Cdh1 or the E2F-inhibitor p105 retinoblastoma protein (pRb). Depleting cells of Emi1 through RNA interference prevents accumulation of cyclin A and inhibits S phase entry. | |
| Store at 4C. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction