missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ EMI 48

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
EMI 48 is an inhibitor of mutant EGFR; it inhibits L858R/T970M/C797S mutants and ex19del/T790M/C797S triple mutants (inhibitory effects at 5 μM) but has no effect on wild-type EGFR. EMI 48 induces EGFR degradation (<5 μM), and inhibits activation of ERK, S6 and AKT.
Specifications
Specifications
| Chemical Name or Material | 7-(Diethylamino)-3-(5-methyl-2-benzoxazolyl)-2 H-1-benzopyran-2-one |
| CAS | 34564-13-1 |
| Quantity | 10 mg |
| Target | EGF Receptor Inhibitors |
| Molecular Formula | C21H20N2O3 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction