missing translation for 'onlineSavingsMsg'
Learn More

EMG1 Antibody (OTI1B8), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18414315 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414315 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414315 Supplier Novus Biologicals Supplier No. NBP271601MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

EMG1 Monoclonal antibody specifically detects EMG1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMG1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1B8
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias 18S rRNA (pseudouridine-N1-)-methyltransferase NEP1, BWCNS, C2F18S rRNA Psi1248 methyltransferase, EMG1 nucleolar protein homolog (S. cerevisiae), essential for mitotic growth 1, FLJ60792, Grcc2f, NEP1EC 2.1.1.-, Nucleolar protein EMG1 homolog, probable ribosome biogenesis protein NEP1, Protein C2f, Ribosome biogenesis protein NEP1
Host Species Mouse
Immunogen Full length human recombinant protein of human EMG1(NP_006322) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 10436
Target Species Human, Mouse, Rat, Canine, Primate, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.