missing translation for 'onlineSavingsMsg'
Learn More

Emetine Dihydrochloride, Mikromol™

Product Code. 18072610
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 18072610

Brand: Mikromol MM0131.02

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Emetine Dihydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Emetine Dihydrochloride
CAS 316-42-7
Analyte or Component Names Emetine Dihydrochloride
Shipping Condition Room Temperature
Molecular Formula C29 H40 N2 O4 . 2 Cl H
Certifications/Compliance ISO 17025
Synonym Emetine Dihydrochloride, (2S,3R,11bS)-3-Ethyl-1,3,4,6,7,11b-hexahydro-9,10-dimethoxy-2-[[(1R)-1,2,3,4-tetrahydro-6,7-dimethoxy-1-isoquinolinyl]methyl]-2H-benzo[a]quinolizine hydrochloride (1:2), 3-Ethyl-1,3,4,6,7,11b-hexahydro-9,10-dimethoxy-2-[(1,2,3,4-tetrahydro-6,7-dimethoxy-1-isoquinolyl)methyl]-2H-benzo[a]quinolizine dihydrochloride, (2S,3R,11bS)-3-Ethyl-1,3,4,6,7,11b-hexahydro-9,10-dimethoxy-2-[[(1R)-1,2,3,4-tetrahydro-6,7-dimethoxy-1-isoquinolinyl]methyl]-2H-benzo[a]quinolizine dihydrochloride, 6',7',10,11-Tetramethoxy-emetan dihydrochloride, Emetine dihydrochloride, (-)-Emetine dihydrochloride, Emetine hydrochloride, Hemometina, l-Emetine dihydrochloride
InChI Formula InChI=1S/C29H40N2O4.2ClH/c1-6-18-17-31-10-8-20-14-27(33-3)29(35-5)16-23(20)25(31)12-21(18)11-24-22-15-28(34-4)26(32-2)13-19(22)7-9-30-24;;/h13-16,18,21,24-25,30H,6-12,17H2,1-5H3;2*1H/t18-,21-,24+,25-;;/m0../s1
SMILES Cl.Cl.CC[C@H]1CN2CCc3cc(OC)c(OC)cc3[C@@H]2C[C@@H]1C[C@H]4NCCc5cc(OC)c(OC)cc45
IUPAC Name (2S,3R,11bS)-2-[[(1R)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-1-yl]methyl]-3-ethyl-9,10-dimethoxy-2,3,4,6,7,11b-hexahydro-1H-benzo[a]quinolizine;dihydrochloride
Molecular Weight (g/mol) 553.56
Quantity 100 mg
Formula Weight 552.2522 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.