missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Emerin Recombinant Rabbit Monoclonal Antibody (23GB5320), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | Emerin |
|---|---|
| Clone | 23GB5320 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30088891
|
Invitrogen™
MA553884 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087532
|
Invitrogen™
MA553885 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Emerin Recombinant Monoclonal antibody specifically detects Emerin in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Emerin | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O08579, P50402, Q63190 | |
| 13726, 2010, 25437 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5320 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AW550900; EDMD; EMD; Emerin; LEM domain containing 5; LEMD5; STA | |
| A synthesized peptide derived from human Emerin (200-254AA). | |
| Primary | |
| -20°C | |
| Emd |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title