missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Emerin Recombinant Rabbit Monoclonal Antibody (10Y5K9)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535459
This item is not returnable.
View return policy
Description
Immunogen sequence: RERPMYGRDS AYQSITHYRP VSASRSSLDL SYYPTSSSTS FMSSSSSSSS WLTRRAIRPE NRAPGAGLGQ DRQVPLWGQL LLFLVFVIVL FFIYHFMQAE EGNPF.
Xeroderma pigmentosum type G (XPG) is a human genetic disease exhibiting extreme sensitivity to sunlight. The XPG protein, a member of the flap endonuclease 1 (FEN-1) structure-specific DNA repair endonuclease family, is an enzyme essential for DNA repair of the major kinds of solar ultraviolet (UV)-induced DNA damages. Human XPG nuclease makes the 3' incision during nucleotide excision repair of DNA. The enzyme cleaves model DNA bubble structures specifically near the junction of unpaired DNA with a duplex region. A 29-amino acid region of human XPG (residues 981-1009) contains the PCNA binding activity. A conserved arginine in XPG (Arg992) is crucial for its PCNA binding activity. Replication Protein A (RPA) binds specifically and directly to two excision repair proteins, the xeroderma pigmentosum damage-recognition protein XPA and the endonuclease XPG.
Specifications
| Emerin | |
| Recombinant Monoclonal | |
| 0.6 mg/mL | |
| PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3 | |
| O08579, P50402, Q63190 | |
| Emd | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 10Y5K9 | |
| Unconjugated | |
| Emd | |
| AW550900; EDMD; EMD; Emerin; LEM domain containing 5; LEMD5; STA | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13726, 2010, 25437 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction