Learn More
Invitrogen™ Emerin Monoclonal Antibody (5A10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527874
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human placenta tissue, human Caco-2 whole cell, human HepG2 whole cell, Rabbit IgG, Marker 1113, human Jurkat whole cellhuman MDA-MB-453 whole cell, human SK-OV-3 whole cell, human SW620 whole cell. IHC: human gastric cancer tissue, human rectal cancer tissue. Flow: A431 cell.
Xeroderma pigmentosum type G (XPG) is a human genetic disease exhibiting extreme sensitivity to sunlight. The XPG protein, a member of the flap endonuclease 1 (FEN-1) structure-specific DNA repair endonuclease family, is an enzyme essential for DNA repair of the major kinds of solar ultraviolet (UV)-induced DNA damages. Human XPG nuclease makes the 3' incision during nucleotide excision repair of DNA. The enzyme cleaves model DNA bubble structures specifically near the junction of unpaired DNA with a duplex region. A 29-amino acid region of human XPG (residues 981-1009) contains the PCNA binding activity. A conserved arginine in XPG (Arg992) is crucial for its PCNA binding activity. Replication Protein A (RPA) binds specifically and directly to two excision repair proteins, the xeroderma pigmentosum damage-recognition protein XPA and the endonuclease XPG.
Specifications
| Emerin | |
| Monoclonal | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| P50402 | |
| Emd | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human Emerin (1-48aa MDNYADLSDTELTTLLRRYNIPHGPVVGSTRRLYEKKIFEYETQRRRL). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 5A10 | |
| Unconjugated | |
| Emd | |
| AW550900; EDMD; EMD; Emerin; LEM domain containing 5; LEMD5; STA | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 2010 | |
| -20°C | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.