missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emeramide, MedChemExpress
Description
Emeramide is a thiol-redox antioxidant and heavy metal chelator.
Specifications
Specifications
| Chemical Name or Material | Emeramide |
| Physical Form | Solid |
| CAS | 351994-94-0 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C12H16N2O2S2 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | BDTH2 |
| Solubility Information | DMSO : 100 mg/mL (351.62 mM; Need ultrasonic and warming) ∣H2O : 1 mg/mL (3.52 mM; Need ultrasonic and warming) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?