missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Luminescence Enhancer For Solution Assays, Emerald-II™ Enhancer

Product Code. 12684315
Click to view available options
Quantity:
2500 assays
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12684315

Brand: Invitrogen™ T2115

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Emerald-II chemiluminescence enhancer significantly reduces aqueous quenching and tunes the wavelength of light emission.

The Emerald-II chemiluminescence enhancer significantly reduces aqueous quenching and tunes the wavelength of light emission.

• Aqueous environments reduce chemiluminescent signal intensity of 1,2-dioxetane substrates by water-induced quenching; chemiluminescence enhancers increase the emission efficiency of light production by partitioning the water from the site of chemiluminescent signal production
• Chemically optimized formulations enhance signal intensity in solution-based applications with dioxetane-based substrate solutions (CSPD™, CDP-Star™, Galacton™, Galacton-Plus™, Galacton-Star™, or NA-Star™ substrates)
• Choice of Emerald-II enhancer facilitates assay development by allowing users to optimize assay signal intensity and wavelength of light emission

Optimize chemiluminescence signals
Emerald-II chemiluminescence enhancer is an essential component of tube or microplate-based assays, significantly reducing the effect of aqueous quenching. Enhancers also tune the wavelength of light emission. Emerald-II enhancer shifts the light emission maximum to 461 nm from 475 nm observed with the dioxetane alone. The use of Emerald-II enhancer can produce a wide dynamic range since photodetector saturation is less likely to occur at higher sample concentrations.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 2500 assays
Format Liquid
Product Line EMERALD-II, NovaBright
Substrate Alkaline Phosphatase
Substrate Type AP (Alkaline Phosphatase) Substrate
Content And Storage Store the product at 4°C.
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.