missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EME1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527110
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EME1 and MUS81 form an endonuclease complex that cleaves branched DNA structures, especially those arising during stalled DNA replication (Abraham et al., 2003).
Specifications
| EME1 | |
| Polyclonal | |
| Unconjugated | |
| EME1 | |
| 6820428D13; Crossover junction endonuclease EME1; EME1; essential meiotic endonuclease 1 homolog 1; essential meiotic endonuclease 1 homolog 1 (S. pombe); essential meiotic endonuclease 1 homolog 2; essential meiotic structure-specific endonuclease 1; hMMS4; homolog of yeast EME1 endonuclease; MMS4; MMS4 homolog; MMS4L; Msm4; SLX2 structure-specific endonuclease subunit homolog A; SLX2A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 146956 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96AY2 | |
| EME1 | |
| Recombinant fragment corresponding to a region within amino acids 1 and 252 of Human EME1 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction