missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EME1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576372
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
EME1 and MUS81 form an endonuclease complex that cleaves branched DNA structures, especially those arising during stalled DNA replication (Abraham et al., 2003).
Specifications
| EME1 | |
| Polyclonal | |
| Unconjugated | |
| EME1 | |
| 6820428D13; Crossover junction endonuclease EME1; EME1; essential meiotic endonuclease 1 homolog 1; essential meiotic endonuclease 1 homolog 1 (S. pombe); essential meiotic endonuclease 1 homolog 2; essential meiotic structure-specific endonuclease 1; hMMS4; homolog of yeast EME1 endonuclease; MMS4; MMS4 homolog; MMS4L; Msm4; SLX2 structure-specific endonuclease subunit homolog A; SLX2A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 146956, 268465, 287634 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8BJW7, Q96AY2 | |
| EME1 | |
| Recombinant fusion protein of human EME1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction