missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EME1 Antibody (MTA31 7h2/1), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50386AF647
This item is not returnable.
View return policy
Description
EME1 Monoclonal specifically detects EME1 in Human samples. It is validated for Western Blot.
Specifications
| EME1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| EME1 | |
| Human EME1 | |
| 0.1 mL | |
| DNA Repair, Homologous Recombination | |
| 146956 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot | |
| MTA31 7h2/1 | |
| Western Blot | |
| crossover junction endonuclease EME1, EC 3.1.22, EC 3.1.22.-, essential meiotic endonuclease 1 homolog 1 (S. pombe), essential meiotic endonuclease 1 homolog 2, FLJ31364, hMMS4, homolog of yeast EME1 endonuclease, MMS4, MMS4 homolog, MMS4L, SLX2 structure-specific endonuclease subunit homolog A, SLX2A | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction