missing translation for 'onlineSavingsMsg'
Learn More

EMD638683 R-Form, MedChemExpress

Product Code. p-200097795
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492467 50 mg 50mg
16456358 10 mg 10mg
16448947 5 mg 5mg
16436666 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16492467 Supplier Medchem Express Supplier No. HY15193A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress EMD638683 R-Form is the R-form of EMD638683. EMD638683 is a highly selective SGK1 inhibitor with IC50 of 3 μM.

EMD638683 R-Form is the R-form of EMD638683. EMD638683 is a highly selective SGK1 inhibitor with IC50 of 3 μM.

Specifications

Chemical Name or Material EMD638683 R-Form
CAS 1184940-47-3
Color Pink
Purity Grade Notes Research
Molecular Formula C18H18F2N2O4
Quantity 50 mg
Solubility Information DMSO : 180 mg/mL (494.04 mM; Need ultrasonic and warming)
SMILES CC1=C(CC)C(C(NNC([C@H](O)C2=CC(F)=CC(F)=C2)=O)=O)=CC=C1O
Molecular Weight (g/mol) 364.34
Formula Weight 364.34
Percent Purity 99.03%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.