missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMD638683, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
EMD638683 is a highly selective SGK1 inhibitor, with an IC50 value of 3 μM.
Specifications
Specifications
| Chemical Name or Material | EMD638683 |
| CAS | 1181770-72-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C18H18F2N2O4 |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 50 mg/mL (137.23 mM) |
| SMILES | CC1=C(CC)C(C(NNC(C(O)C2=CC(F)=CC(F)=C2)=O)=O)=CC=C1O |
| Molecular Weight (g/mol) | 364.34 |
| Formula Weight | 364.34 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion