Learn More
Merck Accessories for ZHE Hazardous Waste System
Product Code. 10101451 Shop All Merck Products
Change view
Click to view available options
Type:
2-Way Valve, 1/8 in. NPTM
Cylinder O-Rings
Luer Fitting
Piston O-Rings, Wiper Seal
Piston with O-Rings, Wiper Seal
Quick-Disconnect Female Swagelok
Quick-Disconnect Male Swagelok
Support Screen 90 mm
Tedlar Bag
Tedlar Bag Adapter
Vent/Relief Valve
ZHE Tight Syringe
Unit Size:
Each
Pack of 10
Pack of 3
Pack of 6
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10101451 Piston with O-Rings, Wiper Seal Each
10768051 2-Way Valve, 1/8 in. NPTM Each
10082143 Cylinder O-Rings Pack of 3
10203451 Piston O-Rings, Wiper Seal Pack of 3
10718441 Luer Fitting Each
10430683 ZHE Tight Syringe Each
10079890 - Pack of 6
10508411 Support Screen 90 mm Each
10708251 Quick-Disconnect Female Swagelok Each
10601872 Tedlar Bag Pack of 10
10602062 Tedlar Bag Adapter Each
10661672 Vent/Relief Valve Each
10718251 Quick-Disconnect Male Swagelok Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 10101451 Supplier Merck Supplier No. YT3009001
Only null left
Add to basket
Add to basket
Accessories for filter holders for sample preparation.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) ZHE Hazardous Waste filtration system
Material Stainless Steel, Fluoroelastomer
Sterility Non-sterile
Type Piston with O-Rings, Wiper Seal