missing translation for 'onlineSavingsMsg'
Learn More

Merck Accessories for ZHE Hazardous Waste System

Accessories for filter holders for sample preparation.

Brand:  Merck YY2209068

208.22 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.



Code : 72

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10079890

  • £224.00 / Pack of 6
Estimated Shipment: 25-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Not Autoclavable
ZHE Hazardous Waste filtration system
Non-sterile
90 mm
O-ring, 2-235, 90mm, fluoroelastomer
Fluoroelastomer
O-Ring, 2-235, 90 mm, Fluoroelastomer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers