missing translation for 'onlineSavingsMsg'
Learn More

Merck Accessories for ZHE Hazardous Waste System

Product Code. 10082143 Shop All Merck Products
Change view
Click to view available options
Type:
2-Way Valve, 1/8 in. NPTM
Cylinder O-Rings
Luer Fitting
Piston O-Rings, Wiper Seal
Piston with O-Rings, Wiper Seal
Quick-Disconnect Female Swagelok
Quick-Disconnect Male Swagelok
Support Screen 90 mm
Tedlar Bag
Tedlar Bag Adapter
Vent/Relief Valve
ZHE Tight Syringe
Unit Size:
Each
Pack of 10
Pack of 3
Pack of 6
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10082143 Cylinder O-Rings Pack of 3
10768051 2-Way Valve, 1/8 in. NPTM Each
10203451 Piston O-Rings, Wiper Seal Pack of 3
10718441 Luer Fitting Each
10430683 ZHE Tight Syringe Each
10079890 - Pack of 6
10508411 Support Screen 90 mm Each
10708251 Quick-Disconnect Female Swagelok Each
10101451 Piston with O-Rings, Wiper Seal Each
10601872 Tedlar Bag Pack of 10
10602062 Tedlar Bag Adapter Each
10661672 Vent/Relief Valve Each
10718251 Quick-Disconnect Male Swagelok Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Product Code. 10082143 Supplier Merck Supplier No. XX6700010
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Accessories for filter holders for sample preparation.

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
For Use With (Equipment) ZHE Hazardous Waste filtration system
Material Fluoroelastomer
Sterility Non-sterile
Type Cylinder O-Rings
Diameter (English) 0.125 in.
Diameter (Metric) Inside 0.3 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.