missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Ultrafiltration Membranes - YM
Shop All Merck Products
Click to view available options
Diameter (Metric):
2.5 cm
4.45 cm
6.35 cm
7.6 cm
Unit Size:
Pack of 10
Specifications
Specifications
| Filter Type | Adsorptive |
| Material (Membrane) | Regenerated Cellulose |
| MWCO | 100 kDa |
| Filter Code | YM-100 |
| Filtration Area | 13.4 cm2 |
| For Use With (Application) | Protein purification, protein concentration, protein desalting |
| Quantity | 10 Pack |
| Type | Ultrafiltration Disc |
| Diameter (English) | 1.75 in. |
| Diameter (Metric) | 4.45 cm |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion