missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
10 kDa NMWL, Ultracel™ regenerated cellulose
Assemble your Amicon Stirred Cell with the membrane of your choice. Ultrafiltration cut disc membranes are available in Ultracel™ regenerated cellulose and Biomax™ polyethersulvone (PES). The membranes are available in a range of different pore sizes & diameters.Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic, tight microstructure of Ultracel membranes assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.Microfiltration Durapore™ PVDF cut discs are available 0.1 μm, 0.22 μm, and 0.45 μm pore sizes in both 63.5 mm and 75 mm diameters.Features & Benefits:•Disc diameters include: 25, 44.5, 47, 63.5, 76, 90 and 150 mm •Both Ultracel™, Durapore™, and Biomax™ membranes are available in a wide range of pore sizes to meet your separation requirements •Compatible with Amicon™ Stirred Cells Applications:Concentration, Buffer Exchange, Desalting
Specifications
Specifications
| Filter Type | Ultrafiltration |
| Material (Membrane) | Regenerated Cellulose |
| MWCO | 10 kDa |
| Filter Code | PLGC |
| Filtration Area | 63 cm2 |
| For Use With (Application) | Protein purification; protein concentration; protein desalting |
| Housing Material | Regenerated Cellulose (RC) |
| Quantity | 5Pack |
| Type | Ultrafiltration Disc |
| Diameter (Metric) Filter | 9 cm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?