missing translation for 'onlineSavingsMsg'
Learn More

Merck Accessories, and Replacement Parts for HP In-Line Stainless Steel Filter Holders 47 mm

Product Code. 10385521 Shop All Merck Products
Change view
Click to view available options
Type:
Filter Holder (High Pressure)
Inner O-Ring
Outer O-Ring
Replacement Parts Kit
Material:
Fluoroelastomer
PTFE
PTFE-treated Fluoroelastomer
Stainless Steel
Unit Size:
Each
Pack of 10
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Material unitSize
10385521 Outer O-Ring Fluoroelastomer Pack of 10
10151921 Inner O-Ring PTFE-treated Fluoroelastomer Pack of 5
10223351 Inner O-Ring PTFE Pack of 5
10577102 Replacement Parts Kit Stainless Steel Each
10339700 Filter Holder (High Pressure) Stainless Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10385521 Supplier Merck Supplier No. XX4504713
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Accessories and parts for filtration of gases or liquids at inlet pressures of 20 to 700 bar.

For filtration of gases or liquids at inlet pressures of 20 to 700 bar

Applications:

  • In-Line Stainless Steel Filter Holders are commonly used for the filtration of gases or liquids at inlet pressures of 20 to 700 bar.
  • Differential pressure depends on filter type used.
  • All filter holders may be autoclaved (available in 25 mm and 47 mm).
  • Filter holder XX4404700 is equipped with a back-pressure support screen.
    • It may be autoclaved with filter in-place, if PTFE-coated support screens, XX4404702 and XX4404704, are used.
  • XX4204709, XX4304701, XX4404703, 5614, and 00907P are replacement parts for Filter Holder XX4404700.
    • Filter holder XX4504700 may be equipped with an upstream filter support screen, XX4504704, for back-pressure protection of filter.
  • XX4504704, XX4504705, XX4504710, XX4504713, and XX45047RK are replacement parts for Filter Holder XX4504700.
  • Replacement parts for the 25 mm SS Filter Holder are available as a kit under Catalog Number XX45025RK, which includes:
    • (3) Stainless Steel Hex-Cap Screws
    • (3) LCR-treated Buna-N O-rings (2-018)
    • (1) Stainless Steel Support Screen
    • (1) Allen™ wrench

SS Filter, Depends on filter type used.

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Disposable Reusable
For Use With (Equipment) 47 mm High Pressure SS Filter Holder
Material Fluoroelastomer
Sterility Non-sterile
Type Outer O-Ring
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.