missing translation for 'onlineSavingsMsg'
Learn More

Merck SNAP i.d.™ 2.0 Protein Detection System

Product Code. p-4507761 Shop All Merck Products
Change view
Click to view available options
Length (English) Gel:
2.95, 5.31 in.
3.3 in.
5.31 in.
Length (Metric) Gel:
13.5 cm
8.4 cm
8.4, 13.5 cm
Width (English) Gel:
2.95 in.
2.95, 3.34 in.
3.34 in.
Width (Metric) Gel:
7.5 cm
7.5, 8.5 cm
8.5 cm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Gel Length (Metric) Gel Width (English) Gel Width (Metric) Gel unitSize
13208319 5.31 in. 13.5 cm 3.34 in. 8.5 cm Each
13298309 3.3 in. 8.4 cm 2.95 in. 7.5 cm Each
13238319 2.95, 5.31 in. 8.4, 13.5 cm 2.95, 3.34 in. 7.5, 8.5 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13208319 Supplier Merck Supplier No. SNAP2MIDI
Add to basket
Add to basket
This item is not returnable. View return policy

Produces blots of a very high quality every time

The fast, versatile SNAP i.d. 2.0 system is designed to fully exploit three-dimensional reagent distribution. Unlike conventional Western blotting, where diffusion is the primary means of reagent transport, the SNAP i.d. 2.0 system applies a vacuum to actively drive antibodies and buffers right through the membrane. This advanced technology promotes antigen binding and thorough washing, enabling you to better optimize your Western blotting conditions.

The EMD Millipore SNAP i.d. 2.0 Protein Detection System shortens time required for blocking, washing and antibody incubations to 30 minutes, allowing you to optimize immunodetection conditions for the highest quality results.

  • Unique vacuum-driven technology and a built-in flow distributor actively drive reagents through the membrane, ensuring even distribution
  • Two blot holder sizes accommodatei Mini (7.5 x 8.4 cm) (Cat. No. SNP2BHMN100) or Midi (8.5 x 13.5cm) (Cat. No. SNP2BHMD100) size gels and two blot holders can be run in parallel

TRUSTED_SUSTAINABILITY

Specifications

Type Protein Detection System Mini
Includes Detection base, 2 midi blot holding frames, 2 antibody collection trays, vacuum tubing, rolling pad and blot roller, 2 wetting trays and a quick start guide
Temperature (Metric) Storage Room Temperature
Length (English) Gel 5.31 in.
Length (Metric) Gel 13.5 cm
Width (English) Gel 3.34 in.
Width (Metric) Gel 8.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.