Learn More
Merck SNAP i.d. 2.0™ IHC Frame
For use with SNAP i.d. 2.0 protein detection system
Brand: Merck SNAP2FRIHC
Additional Details : Weight : 0.10000kg
Includes: Incubation cover and assembly fixture
Description
- Enables to process up to 12 tissue slides
- For use with immunohistochemistry slides
Specifications
Incubation cover and assembly fixture | |
1/Pk. |