missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck RNase A
Specifications
Specifications
| For Use With (Application) | Used as a reagent with montage plasmid miniprepHTS 96 kit for high-purity plasmid or Bacterial Artificial Chromosome minipreps |
| Content And Storage | 15°C to 30°C |
| Quantity | 0.9 mL |
| Product Type | Ribonuclease A |
Safety and Handling
Product Identifier
- RNase A Reagent
- Danger
- Respiratory sensitiser Category 1
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: ribonuclease
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?