missing translation for 'onlineSavingsMsg'
Learn More

Merck Replacement Solutions for Montage™ BAC96 Miniprep Kit and Montage Plasmid Miniprep96 Kit

Suitable for cell resuspension for plasmid & BAC minipreps

Brand:  Merck LSKNS0500

Code : NEW

Additional Details : Weight : 0.36800kg Transport : UN number : 2790 Chem class : 8 Pack group : III

 View more versions of this product

Product Code. 11704684

  • £166.00 / 500mL
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Montage Plasmid MiniprepHTS 96 Kit is a fast, easy-to-use kit for high-purity plasmid or bacterial artificial chromosome (BAC) minipreps
Specifications

Specifications

Plasmid Miniprep, BAC Prep
Neutralization Solution
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers