missing translation for 'onlineSavingsMsg'
Learn More

Merck ReBlot Plus Kit and Reagents

Product Code. 10106803
Change view
Click to view available options
Quantity:
50 mL
Ea.
Product Type:
Mild WB Stripping Buffer
Strong WB Stripping Buffer
Western Blot Kit
Unit Size:
1 set
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type unitSize
10106803 50 mL Mild WB Stripping Buffer 50mL
11360250 Ea. Western Blot Kit 1 set
10721814 50 mL Strong WB Stripping Buffer 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Please note: We no longer supply Millipore Antibodies.

Product Code. 10106803 Supplier Merck Supplier No. 2502

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Please note: We no longer supply Millipore Antibodies.

Includes

Mild Antibody Stripping Solution (10X), 50mL; Strong Antibody Stripping Solution (10X), 50mL

Blots can be stripped of antibodies in approximately 15 minutes at room temperature

Western blotting is a commonly used technique for studying protein function and localization. Typically, protein samples are electrophoresed on SDS-PAGE and transferred to a membrane such as nitrocellulose or nylon, where they are probed with specific antibodies. Unlike nucleic acid-based technologies, which allow reuse of Southern and Northern blots, it has been difficult to reuse Western blots.

  • No pungent-smelling β-mercaptoethanol is contained in the Antibody Stripping Solution
  • Antibody stripping is done at room temperature—no heating of blots is required
  • Blots may be reused in 25 min.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Mild WB Stripping Buffer
For Use With (Application) Western Blot
Quantity 50 mL
Concentration 2X
Kit Contents 25mL Mild Antibody stripping solution 10X
Content And Storage 2°C to 8°C
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Re-Blot Plus Mild Solution (10X)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308-IF exposed or concerned:.
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide, sodium azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.