missing translation for 'onlineSavingsMsg'
Learn More

Merck PureProteome™ Human Albumin/Immunoglobulin Depletion Kit

Product Code. 15385442
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15385442

Brand: Merck LSKMAGHDKIT

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PureProteome™ magnetic beads are a powerful system to isolate antibodies faster, easier and with better reproducibly then ever before.

The PureProteome™ human albumin/immunoglobulin depletion kit is a magnetic bead based kit that enables high depletion efficiency (typically >99%) of albumin and all immunoglobulins (i.e IgG, IgA, IgM, IgE and IgD) from human serum or plasma samples. The kit includes 12ml of premixed mag beads, 7ml of PBS buffer and an 8 pack of Amicon Ultra centrifugals for downstream buffer exchange or concentration if required.

  • High capacity: Advanced chemistry combined with high surface area provides more binding sites for proteins
  • Consistent results with no sample loss: Particles visible as they adhere to side of tube for quick and easy aspiration and complete buffer removal
  • Fast processing time: Beads are immobilized in seconds and increased kinetics of bead-protein binding enables shorter incubations
  • Economical: Significantly more affordable then competitive magnetic beads

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Human serum/plasma depletion

TRUSTED_SUSTAINABILITY

Specifications

Cross Reactivity Human
Includes 12mL Premixed magnetic beads, 7mL of PBS Buffer, 8 Pack Amicon Ultra 2mL Centrifugal filters, 3K MWCO
Quantity 1 Kit
Sample Type Serum, Plasma
Sample Volume 12 mL
Content And Storage 2°C to 8°C
For Use With (Application) Human Serum, Plasma depletion
Product Type PureProteome™ Human Albumin/Immunoglobulin depletion kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.