Learn More
Merck pRSF-1b DNA Vector
Product Code. 16854650 Shop All Merck Products
Change view
Click to view available options
Quantity:
10 μg
Product Code. Quantity
16854650 10 μg
1 options
Product Code. 16854650 Supplier Merck Supplier No. 713633
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The pRSF-1b plasmid features coexpression capabilities as well as the ability to express fusion proteins with a N-terminal His·Tag™ coding sequence that results in native protein after purification and cleavage. This plasmid carries an origin derived from RSF1030 (1, 2) and kanamycin resistance, allowing for the option of coexpression with many other Novagen T7-based expression vectors. This plasmid contains a strong T7lac promoter, an amino-terminal His·Tag coding sequence, and multiple cloning site (MCS) regions designed to allow the generation of target proteins with minimal vector encoded fusion.
TRUSTED_SUSTAINABILITY
Specifications
Marker Gene Kanamycin resistance
Promoter T7lac
Content And Storage Store at or below -70°C.