Learn More
Merck Pipe Thread Fittings
Product Code. p-3303595 Shop All Merck Products
Click to view available options
Type:
Pipe Thread Fitting
Pipe Thread Valve
Connection Type:
1/4 in. NPTM
1/4 in. NPTM Close Nipple
1/8 in. -1/8 in. NPTM 3-way Plug Valve
Unit Size:
Each
Product Code. 10410493
Brand: Merck XX6700101
Only null left
Add to basket
Add to basket
Use in conjunction with filtration systems.
Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.Tri-Clamp (TC) fittings are referred to by convention as 1 1/2 in., 2 in., etc. 1 1/2 in. TC fittings connect to 1 in. or 1 1/2 in. O.D. tube sizes. 2 in. TC fittings connect to 2 in. O.D. tube size only.
TRUSTED_SUSTAINABILITY
Specifications
Connection Type 1/4 in. NPTM
For Use With (Equipment) Water Purification System
Type Pipe Thread Fitting
Material Series 300 stainless steel
Diameter (English) Inside 0.25 in.
Diameter (Metric) Inside 0.63 cm