missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Includes
Operating instructions and Certificate of Quality
Designed for process development and sample preparation, with an optimal processing volume range of 50 to 1000mL.
- True linear scalability with same flow path, channel length, and channel height as other established Pellicon products
- Can operate with peristaltic pump or Labscale™ TFF system Robust thermoplastic (polypropylene) construction
- Controlled channel height
- Choice of Biomax™ PES (polyethersulfone) or Ultracel™ PLC (composite regenerated cellulose) membranes
Concentration and desalting of proteins, buffer exchange, depyrogenation, clarification, and preparation of material for clinical trials.
Order Info
Packaged one per box.
Specifications
Specifications
| Filter Type | Ultrafiltration |
| Format | Cassette |
| Material | Polypropylene |
| MWCO | 10 kDa |
| Pore Size | 100 μm |
| Filtration Area | 50 cm2 |
| For Use With (Application) | Protein purification, analytical sample preparation |
| Includes | Operating instructions and certificate of quality |
| Max. Inlet Pressure | 80 psi at 50°C |
| pH Range | 2 to 21 |
| Show More |
Safety and Handling
Product Identifier
- PELXL PL10K C 50CM2
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- MIXTURE LIST-Contain: phosphoric acid, acetic acid
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?