missing translation for 'onlineSavingsMsg'
Learn More

Merck Pellicon™ XL Devices

Product Code. 15789048 Shop All Merck Products
Change view
Click to view available options
MWCO:
10 kDa
30 kDa
300 kDa
5 kDa
50 kDa
pH Range:
1 to 14
2 to 21
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO pH Range unitSize
15789048 300 kDa 1 to 14 Each
11943926 5 kDa 1 to 14 Each
10184739 10 kDa 1 to 14 Each
12170212 30 kDa 1 to 14 Each
11913916 50 kDa 1 to 14 Each
13197494 30 kDa 2 to 21 Each
12801666 10 kDa 2 to 21 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15789048 Supplier Merck Supplier No. PXB300C50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Operating instructions and Certificate of Quality

Small-volume tangential flow filter device

Designed for process development and sample preparation, with an optimal processing volume range of 50 to 1000mL.

  • True linear scalability with same flow path, channel length, and channel height as other established Pellicon products
  • Can operate with peristaltic pump or Labscale™ TFF system Robust thermoplastic (polypropylene) construction
  • Controlled channel height
  • Choice of Biomax™ PES (polyethersulfone) or Ultracel™ PLC (composite regenerated cellulose) membranes

Concentration and desalting of proteins, buffer exchange, depyrogenation, clarification, and preparation of material for clinical trials.

Order Info

Packaged one per box.

TRUSTED_SUSTAINABILITY

Specifications

Filter Type Ultrafiltration
Format Cassette
Material Polypropylene
MWCO 300 kDa
Pore Size 100 μm
Filtration Area 50 cm2
For Use With (Application) Protein purification, analytical sample preparation
Includes Operating instructions and certificate of quality
Max. Inlet Pressure 80 psi at 50°C
pH Range 1 to 14
Quantity 1 Pack
Recirculation Rate 0.03 to 0.05L/min.
Type Pellicon XL Ultrafiltration Device
Volume (English) Working 0.5 oz.
Volume (Metric) Hold Up 3.2 mL
Volume (Metric) Working 15 mL
Dimensions (L x W) 7.4 x 1.2 in. (18.8 x 3cm)
Length (English) 7.4 in.
Length (Metric) 18.8 cm
Width (English) 1.2 in.
Width (Metric) 3 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.