missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore Nutrient Media

Product Code. 17538046
Change view
Click to view available options
Media Type:
mFC Fecal Broth
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Media Type unitSize
17538046 mFC Fecal Broth Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17538046 Supplier Merck Millipore Supplier No. MHA000P2F

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Developed specifically for culturing microorganisms on membrane filters; select media type to match organism type and analytical test needs

  • Each dry powder lot for media preparation assayed for purity and compared to satisfactory lot
  • Each m-Endo broth for total coliform lot and m-FC broth for fecal coliform analysis lot certified in accordance with the Safe Drinking Water Act
  • Lot QC certificates included in each package as added assurance for critical procedures

Order Info

Nutrient media comes in liquid form, packaged in 2mL plastic ampules. Refrigerate as detailed on label.

TRUSTED_SUSTAINABILITY

Specifications

Color Deep Red
Content And Storage 2°C to 10°C
Form Liquid
Manufacturing Origin United States
Media Type mFC Fecal Broth
Product Type Microbiology Media
Target Organism Class Coliforms
For Use With (Application) Water testing for Coliforms
Packaging Type Ampule
Quantity 50 x 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.