Learn More
Merck MultiScreen™ Multiple Punch Kits
Product Code. 11566365 Shop All Merck Products
Change view
Click to view available options
For Use With (Equipment):
12 x 75 mm Tubes
4 mL Vials
7 mL Vials
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11566365 4 mL Vials Each
11556365 7 mL Vials Each
11722594 12 x 75 mm Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11566365 Supplier Merck Supplier No. MAPK8960B
Only null left
Add to basket
Add to basket
Includes punch with punch carrier plate and punch distributor
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 4 mL Vials
For Use With (Application) Radionucleotide counting by transfer of individual filters to tubes
Includes Multiple punch, 2 carrier racks for 4mL vials
Quantity 1/Pk.