missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
96-well plate and 96-well receiver tray fit together to provide a complete system for culturing, testing, and reading assay results in a one-step protocol. Simply transfer parasites, nematodes or other multicellular organisms into the wells, treat with target compound, and measure the migration of the organisms.
- Provide a ready-to-use system for assays measuring paralysis, cytotoxicity, and death of multicellular organisms
- Optically clear for use with an imaging microscope or other microscopic analysis
- Can be coated with a thin layer of agar if necessary
- Have also been shown to be useful as high-throughput cell strainers prior to cellular analysis in a FACS™ analyzer
Specifications
Specifications
| Format | Standard |
| Membrane | Nylon Mesh |
| Array | 8 x 12 |
| Pore Size | 40 μm |
| Color | Clear |
| Material | Styrene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Filtration Area | 0.28 cm2 |
| For Use With (Application) | Whole organism screening |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?