missing translation for 'onlineSavingsMsg'
Learn More

Merck MultiScreen™ HTS 96-Well Filter Plates with MCE Membrane

Commonly used for Elispot assays, bacterial screening, ELISA or viral infection studies

Brand:  Merck MSHAN4B50

Code : 73

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10307183

  • £1940.00 / Pack of 50
Estimated Shipment: 10-09-2024
to see stock.
Add to basket

Description

Description

  • Developed for high-throughput use with automated work stations
  • Individually sealed wells prevent incubation crosstalk
  • Removable underdrain allows access to the filters
  • Rigid sidewalls improve handling
  • Ample surfaces provided for bar code labels
  • Pore size: .45μm

Solid-phase RIA, DNA hybridization, ELISA and protein binding

Specifications

Specifications

Standard
8 x 12
Opaque
96
0.28cm2
50/Pk.
Mixed Cellulose Esters
0.45 μm
Barex
Non-sterile
Solid phase RIA, DNA hybridization, ELISA, protein binding
8.55 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Promotions

Promotions