missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Developed for high-throughput use with automated work stations
- Individually sealed wells prevent incubation crosstalk
- Removable underdrain allows access to the filters
- Rigid sidewalls improve handling
- Ample surfaces provided for bar code labels
- Pore size: .45μm
Solid-phase RIA, DNA hybridization, ELISA and protein binding
Specifications
Specifications
| Format | Standard |
| Membrane | Mixed Cellulose Esters |
| Array | 8 x 12 |
| Pore Size | 0.45 μm |
| Color | Clear |
| Product Type | MultiScreenHTS-HA Filter Plate |
| Material | Styrene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Filtration Area | 0.28 cm2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?