missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreenHTS 384-Well Filter Plates
Use in automated, high throughput applications, including biological assays and sample preparation. MultiScreenHTS 384-Well Filter Plates are non-sterile, single use plates available in several mesh materials and pore sizes to meet the needs of different applications.
Brand: Merck MZFCN0W50
This item is not returnable.
View return policy
Description
For use in receptor-ligand binding, general sample preparation, enzyme assays with precipitation and DNA purification.- Rigid sidewalls allows full compatibility with automated/robotic systems
- Unique plate design and individually sealed filter discs prevent cross-talk between wells
- Opaque plastic lids prevents light transfer between wells during direct scintillation counting
- Barcoding available
- Meet ANSI™/SBS standards
Specifications
Standard | |
16 x 24 | |
Opaque, White Bottom | |
384 | |
0.09 cm2 | |
50/Pk. | |
1.28 cm | |
1.44 cm |
Glass Fiber, Polyester Mesh | |
1.2 μm | |
Barex | |
Non-sterile | |
Receptor, ligand binding assays, sample prep, TCA precipitation | |
1.28 x 8.55 x 1.44 cm | |
8.55 cm |