Learn More
Merck MultiScreen™ Column Loaders
Product Code. 10208490 Shop All Merck Products
Change view
Click to view available options
Volume (Metric) Well:
25 μL
45 μL
100 μL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Well unitSize
10208490 100 μL Each
10760113 25 μL Each
10196670 45 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10208490 Supplier Merck Supplier No. MACL09600
Only null left
Add to basket
Add to basket
No need to pipet slurries, gels, or resins; load all 96 wells simultaneously and uniformly
  • Constructed of aluminum with an abrasion-resistant, chemically inert, nonstick coating
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Product Type MultiScreen Column Loader
Volume (Metric) Well 100 μL
Sterility Non-sterile
For Use With (Application) Load 100μL resin volumes simultaneously into 96 wells of MultiScreen plate for resin or bead based assays, chromatography
Quantity 1/Pk.