missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Constructed of aluminum with an abrasion-resistant, chemically inert, nonstick coating
Specifications
Specifications
| Format | Standard |
| Color | Black |
| Volume (Metric) Well | 45 μL |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| For Use With (Application) | Load 45μL resin volumes simultaneously into 96 wells of MultiScreen plate for resin or bead based assays, chromatography |
| Quantity | 1/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?