missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreen™ 96-Well, Non-Treated, Flat-Bottom Microplate
Color:Clear; Sterility:Sterile; Lid:Without Lid
Brand: Merck MAMCS9610
Code : 71
Additional Details : Weight : 0.10000kg
Description
For basolateral feeding and/or co-culture in a single well or 96-well format
- Use with sterilized MultiScreen filter plates
- Polystyrene with cut-off corners
- Sterilized and individually wrapped
- Untreated surfaces
- Independent wells fit the 96 wells of the membrane plate
- Patented “tear drop” shaped wells prevent air entrapment under membrane
Specifications
Standard | |
8 x 12 | |
Polystyrene | |
Sterile | |
Flat Bottom (F-well) | |
10/Pk. |
Without Lid | |
Clear | |
96 | |
Non-treated | |
Cell transport assays, use with sterile MultiScreen filtration plate |