missing translation for 'onlineSavingsMsg'
Learn More

Merck MultiScreen™ Cell Culture Trays

Product Code. 10340271 Shop All Merck Products
Change view
Click to view available options
Well Count:
1
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Well Count unitSize
10340271 1 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10340271 Supplier Merck Supplier No. MAMCS0110

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For basolateral feeding and/or co-culture in a single well or 96-well format

  • Use with sterilized MultiScreen filter plates
  • Polystyrene with cut-off corners
  • Sterilized and individually wrapped
  • Untreated surfaces
Single-Well Tray
  • Holds medium and commonly feeds all 96 wells
  • 96-Well Tray
    • Independent wells fit the 96 wells of the membrane plate
    • Patented “tear drop” shaped wells prevent air entrapment under membrane

    TRUSTED_SUSTAINABILITY

    Specifications

    Closure Included Closure Not Included
    Bottom Shape Flat
    Color Clear
    Material Styrene
    Shape Rectangular
    Sterility Sterile
    Surface Treatment Non-treated
    Type Cell Culture Dish
    Well Count 1
    For Use With (Application) Cell culture applications, use with sterile MultiScreen filtration plate
    Packaging Individually Wrapped
    Quantity 10/Pk.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.